Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Platinium(IV) oxide hydrate Pt 79-84%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

AC21192-0010 AC21192-0050 AC21192-0250
TS21192-0010EA 162 USD InStock
TS21192-0010 TS21192-0050 TS21192-0250
Platinium(IV) oxide hydrate Pt 79-84%
Platinium(IV) oxide hydrate


Formula: O₂Pt·H₂O
MW: 245.09
Melting Pt: 450
Density: 10.2
Storage Temperature: Ambient
MDL Number: MFCD00066964
CAS Number: 52785-06-5
EINECS: 215-223-0
UN: 1479
ADR: 5.1 I
Merck Index: 12,07686
Order Now
Specification Test Results
Appearance Brown to black Powder
Assay 79 to 84 % (Pt)
Surface area ≥90 m²/g (BET)

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty