Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium tetrachloroplatinate(II) 99.99% (trace metals basis)

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

AC43703-0010 AC43703-0050
TS43703-0010EA 167 USD InStock
TS43703-0010 TS43703-0050
Potassium tetrachloroplatinate(II) 99.99% (trace metals basis)
Potassium tetrachloroplatinate(II)


Formula: Cl₄K₂Pt
MW: 415.09
Melting Pt: 265 (dec.)
Density: 3.38
Storage Temperature: Ambient
MDL Number: MFCD00011378
CAS Number: 10025-99-7
UN: 3288
ADR: 6.1 III
Merck Index: 12,07855
Order Now
Specification Test Results
Appearance Orange to red to brown Crystals
Total trace metal impurities ≤100 ppm

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty