Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethylaluminium chloride 0.9 M in heptane, AcroSeal™

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

AC37779-1000 AC37779-8000
TS37779-1000EA 104 USD InStock
TS37779-1000 TS37779-8000
Dimethylaluminium chloride 0.9 M in heptane, AcroSeal™
Dimethylaluminium chloride


Formula: C₂H₆AlCl
Flash Pt: –4 °C
Storage Temperature: Ambient
CAS Number: 1184-58-3
EINECS: 214-668-8
UN: 3399
ADR: 4.3,I
Order Now
Specification Test Results
Appearance Clear to slightly hazy colorless Solution (May develop some turbidity or precipitate)
Titration Argentometric 0.85 to 1.00 M

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty