Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tetraethylammonium fluoride hydrate 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

AC21666-0050 AC21666-0250
TS21666-0050EA 52 USD InStock
TS21666-0050 TS21666-0250
Tetraethylammonium fluoride hydrate 98%
Tetraethylammonium fluoride hydrate


Formula: C₈H₂₀FN·xH₂O
MW: 149.25 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00149990
CAS Number: 98330-04-2
EINECS: 211-559-7
Order Now
Specification Test Results
Appearance White to beige Crystals or adhering crystals or crystalline solid
Infrared spectrum Conforms
Water ≤28 % (K.F.)
Titration with HClO4 ≥97.5 % (On dry substance)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty