Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bismuth sodium trioxide 85%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

AC20521-0010 AC20521-0250 AC20521-2500
TS20521-0010EA 503 USD InStock
TS20521-0010 TS20521-0250 TS20521-2500
Bismuth sodium trioxide 85%
Bismuth sodium trioxide


Formula: BiNaO₃
MW: 279.97 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003474
CAS Number: 12232-99-4
EINECS: 235-455-6
Merck Index: 12,08728
Order Now
Specification Test Results
Appearance Yellow to brown Powder
Titration Iodometric ≥80 %
Chloride (Cl) ≤20 ppm
Nitrate (NO3) ≤40 ppm
Manganese (Mn) ≤5 ppm

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty