Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bismuth sodium trioxide ACS

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

AC44813-0250 AC44813-1000
TS44813-0250EA 100 USD InStock
TS44813-0250 TS44813-1000
Bismuth sodium trioxide ACS
Bismuth sodium trioxide


Formula: BiNaO₃
MW: 279.97 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003474
CAS Number: 12232-99-4
Merck Index: 12,08728
Order Now
Specification Test Results
Appearance Yellow to brown Solid
Titration with KMnO4 ≥80.0 %
Chloride (Cl) ≤0.002 %
Manganese (Mn) ≤5 ppm
Oxidizing efficiency ≥99.6 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty