Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium molybdate(VI) 95%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

AC39323-1000 AC39323-5000
TS39323-1000EA 79 USD InStock
TS39323-1000 TS39323-5000
Potassium molybdate(VI) 95%
Potassium molybdate(VI)


Formula: K₂MoO₄
Melting Pt: 919 °C
Storage Temperature: Ambient
MDL Number: MFCD00011370
CAS Number: 13446-49-6
Order Now
Specification Test Results
Appearance White Powder and/or chunks
Assay ≥94 %
Chloride (Cl) ≤0.05 %
Phosphate (PO4) ≤0.03 %
Sulfate (SO4) ≤0.05 %
Iron (Fe) ≤0.01 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty