Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tetrapropylammonium chloride 94%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

AC42071-0250 AC42071-1000
TS42071-0250EA 89 USD InStock
TS42071-0250 TS42071-1000
Tetrapropylammonium chloride 94%
Tetrapropylammonium chloride


Formula: C₁₂H₂₈ClN
MW: 221.81
Melting Pt: 230
Density: 1.0334
Storage Temperature: Ambient
MDL Number: MFCD00038729
CAS Number: 5810-42-4
EINECS: 227-375-5
Order Now
Specification Test Results
Appearance White to light yellow Crystals or crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥92 %
Specific optical rotation +33° to +36° (20°C, 589 nm) (c=10, 2N HCl)
Titration with HClO4 ≥98.5 % (On dry substance)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty