Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tetrabutylammonium perchlorate 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

AC42014-1000 AC42014-5000
TS42014-0250EA 108 USD InStock
TS42014-0250 TS42014-1000 TS42014-5000
Tetrabutylammonium perchlorate 98%
Tetrabutylammonium perchlorate


Formula: C₁₆H₃₆ClNO₄
Melting Pt: 210 °C
Storage Temperature: Ambient
MDL Number: MFCD00038722
CAS Number: 1923-70-2
EINECS: 217-655-5
UN: 1479
ADR: 5.1,II
Order Now
Specification Test Results
Appearance White Crystalline powder
Infrared spectrum Conforms
Titration ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty