Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Copper(II) carbonate basic Cu 54-56%, pure

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

AC19667-0025 AC19667-0250 AC19667-5000
TS19667-0025EA 230 USD InStock
TS19667-0025 TS19667-0250 TS19667-5000
Copper(II) carbonate basic Cu 54-56%, pure
Copper(II) carbonate basic


Formula: CuCO₃·Cu(OH)₂
MW: 221.12 g/mol
Melting Pt: 200
Storage Temperature: Ambient
MDL Number: MFCD00010976
CAS Number: 12069-69-1
UN: 3077
ADR: 9 III
Merck Index: 13,02658
Order Now
Specification Test Results
Appearance Green Crystalline powder
Assay 54 to 56 % (Cu)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty