Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Palladium(II) oxide 99.995% (trace metals basis)

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

AC19368-0010 AC19368-0050
TS19368-0010EA 288 USD InStock
TS19368-0010 TS19368-0050
Palladium(II) oxide 99.995% (trace metals basis)
Palladium(II) oxide


Formula: OPd
MW: 122.42 g/mol
Melting Pt: 870 (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011172
CAS Number: 1314-08-5
Merck Index: 12,07125
Order Now
Specification Test Results
Appearance Gray to black Powder or granules and/or chunks
Assay 85.2 to 88.7 % (Pd)
Total trace metal impurities ≤50 ppm

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty