Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Trimethylsilyl)ethanol 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

AC21048-0100 AC21048-0500 AC21048-2500
TS21048-0100EA 146 USD InStock
TS21048-0100 TS21048-0500 TS21048-2500
2-(Trimethylsilyl)ethanol 99%
2-(Trimethylsilyl)ethanol


Formula: HOH₂CCH₂Si(CH₃)₃
Boiling Pt: 71…73 °C (35 mmHg)
Density: 0.825 g/cm³
Flash Pt: 50 °C
Storage Temperature: Ambient
MDL Number: MFCD00002825
CAS Number: 2916-68-9
EINECS: 220-844-5
UN: 1987
ADR: 3,III
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4235 to 1.4255 (20°C, 589 nm)
GC ≥98.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty