Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Allyltrimethylsilane 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

AC34999-0100 AC34999-0500
TS34999-0100EA 57 USD InStock
TS34999-0100 TS34999-0500
Allyltrimethylsilane 97%
Allyltrimethylsilane


Formula: H₂CCHCH₂Si(CH₃)₃
Boiling Pt: 84 °C (1013 hPa)
Density: 0.719 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00008635
CAS Number: 762-72-1
EINECS: 212-104-5
UN: 1993
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty