Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethoxytrimethylsilane 95%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

AC31382-0050 AC31382-0250 AC31382-1000
TS31382-0050EA 17 USD InStock
TS31382-0050 TS31382-0250 TS31382-1000
Ethoxytrimethylsilane 95%
Ethoxytrimethylsilane


Formula: C₅H₁₄OSi
MW: 118.25 g/mol
Boiling Pt: 75
Melting Pt: -83
Density: 0.75 g/cm³
Flash Pt: –18 °C
Storage Temperature: Ambient
MDL Number: MFCD00009069
CAS Number: 1825-62-3
EINECS: 217-370-6
UN: 1993
ADR: 3 II
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3730 to 1.3750 (20°C, 589 nm)
GC ≥94 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty