Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N,O-Bis(trimethylsilyl)trifluoroacetamide with 1% trimethylsilyl chloride

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

AC38932-0250 AC38932-1000 AC38932-5000
TS38932-0250EA 160 USD InStock
TS38932-0250 TS38932-1000 TS38932-5000
N,O-Bis(trimethylsilyl)trifluoroacetamide with 1% trimethylsilyl chloride
N,O-Bis(trimethylsilyl)trifluoroacetamide


Formula: C₈H₁₈F₃NOSi₂
Boiling Pt: 145…147 °C (760 mmHg)
Density: 0.974 g/cm³
Flash Pt: 34 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008269
CAS Number: 25561-30-2
EINECS: 247-103-9
UN: 2920
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
GC ≤1.5 % TMCS

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty