Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Trimethylsulphoxonium iodide 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

AC14026-1000 AC14026-5000
TS14026-1000EA 58 USD InStock
TS14026-1000 TS14026-5000
Trimethylsulphoxonium iodide 98%
Trimethylsulphoxonium iodide


Formula: C₃H₉IOS
MW: 220.07 g/mol
Melting Pt: 175 °C (dec.)
MDL Number: MFCD00011899
CAS Number: 1774-47-6
EINECS: 217-204-2
Order Now
Specification Test Results
Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥97.5 %
Specific optical rotation -142° to -146° (20°C, 589 nm) (c=1, ethanol)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty