Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium chlorodifluoroacetate 96%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

AC40459-0050 AC40459-0250 AC40459-1000
TS40459-0050EA 43 USD InStock
TS40459-0050 TS40459-0250 TS40459-1000
Sodium chlorodifluoroacetate 96%
Sodium chlorodifluoroacetate


Formula: C₂ClF₂NaO₂
Melting Pt: 195…201 °C
Storage Temperature: Ambient
MDL Number: MFCD00064771
CAS Number: 1895-39-2
EINECS: 217-586-0
Order Now
Specification Test Results
Appearance White to off-white Powder
Infrared spectrum Conforms
Titration with HClO4 ≥95 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty