Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethoxy(dimethyl)silane 95+%, AcroSeal™

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

AC35153-0010 AC35153-1000
TS35153-0010EA 142 USD InStock
TS35153-0010 TS35153-1000
Dimethoxy(dimethyl)silane 95+%, AcroSeal™
Dimethoxy(dimethyl)silane


Formula: C₄H₁₂O₂Si
MW: 120.22 g/mol
Boiling Pt: 82
Melting Pt: -80
Density: 0.865 g/cm³
Flash Pt: –8 °C
Storage Temperature: Ambient
MDL Number: MFCD00025691
CAS Number: 1112-39-6
EINECS: 214-189-4
UN: 1993
ADR: 3 II
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3690 to 1.3710 (20°C, 589 nm)
GC ≥95.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty