Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Lanthanum(III) chloride hydrate 64,5 - 70%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

AC19915-0025 AC19915-1000 AC19915-5000
TS19915-0025EA 1169 USD InStock
TS19915-0025 TS19915-1000 TS19915-5000
Lanthanum(III) chloride hydrate 64,5 - 70%
Lanthanum(III) chloride hydrate


Formula: Cl₃La·H₂O
MW: 263.28
Boiling Pt: 1000
Melting Pt: 96
Storage Temperature: Ambient
MDL Number: MFCD00149753
CAS Number: 20211-76-1
EINECS: 233-237-5
UN: 1759
ADR: 8 III
Order Now
Specification Test Results
Appearance White Adhering crystals
Gravimetry 64.5 to 70 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty