Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Picolyl bromide hydrobromide 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

AC39714-0010 AC39714-0050
TS39714-0010EA 50 USD InStock
TS39714-0010 TS39714-0050
3-Picolyl bromide hydrobromide 97%
3-Picolyl bromide hydrobromide


Formula: C₆H₇Br₂N
Storage Temperature: Ambient
MDL Number: MFCD01321314
CAS Number: 4916-55-6
UN: 2923
Order Now
Specification Test Results
Appearance White to yellow to brown Powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 148 to 154 °C
Titration with HClO4 ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty