Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Aluminium trifluoromethanesulphonate 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

AC38326-0100 AC38326-0500
TS38326-0100EA 85 USD InStock
TS38326-0100 TS38326-0500
Aluminium trifluoromethanesulphonate 99%
Aluminium trifluoromethanesulphonate


Formula: C₃AlF₉O₉S₃
MW: 474.19 g/mol
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00143596
CAS Number: 74974-61-1
UN: 3261
ADR: 8 III
Order Now
Specification Test Results
Appearance White Powder
Infrared spectrum Conforms
Titration Complexometric ≥98.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty