Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dichlorodiethylsilane 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

AC33810-0050 AC33810-0250
TS33810-0050EA 48 USD InStock
TS33810-0050 TS33810-0250
Dichlorodiethylsilane 97%
Dichlorodiethylsilane


Formula: C₄H₁₀Cl₂Si
MW: 157.11 g/mol
Melting Pt: –96 °C
Flash Pt: 28 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000493
CAS Number: 1719-53-5
EINECS: 217-005-0
UN: 1767
ADR: 8 II
Order Now
Specification Test Results
Appearance Clear colorless to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.4290 to 1.4320 (20°C, 589 nm)
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty