Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tributylborane 1 M in THF, AcroSeal™

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

AC17710-1000 AC17710-8000
TS17710-1000EA 148 USD InStock
TS17710-1000 TS17710-8000
Tributylborane 1 M in THF, AcroSeal™
Tributylborane


Formula: C₁₂H₂₇B
Density: 0.847 g/cm³
Flash Pt: –20 °C
Storage Temperature: Ambient
MDL Number: MFCD00009423
CAS Number: 122-56-5
EINECS: 204-554-6
UN: 3392
ADR: 4.2,
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Solution
Strength 0.95 to 1.10 M
Loss on drying ≤0.5 % (105°C, 4 hr)
Residue after ignition ≤0.1 %
Titration with HClO4 99.0 to 101.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty