Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Trichlorotris(tetrahydrofuran)titanium(III) 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

AC37516-0010 AC37516-0100
TS37516-0010EA 46 USD InStock
TS37516-0010 TS37516-0100
Trichlorotris(tetrahydrofuran)titanium(III) 97%
Trichlorotris(tetrahydrofuran)titanium(III)


Formula: C₁₂H₂₄Cl₃O₃Ti
Melting Pt: >250 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01631276
CAS Number: 18039-90-2
UN: 2925
ADR: 4.1,II
Order Now
Specification Test Results
Appearance Blue-green to blue Powder or crystals
Infrared spectrum Conforms
Titration Argentometric ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty