Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tris(trimethylsilyl)borate

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

AC38124-0250 AC38124-1000
TS38124-0250EA 114 USD InStock
TS38124-0250 TS38124-1000
Tris(trimethylsilyl)borate
Tris(trimethylsilyl) borate


Formula: C₉H₂₇BO₃Si₃
MW: 278.376 g/mol
Boiling Pt: 183…185 °C (760 mmHg)
Density: 0.834 g/cm³
Flash Pt: 42 °C
Storage Temperature: Ambient
MDL Number: MFCD00051588
CAS Number: 4325-85-3
UN: 1993
ADR: 3 III
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3850 to 1.3870 (20°C, 589 nm)
GC ≥97.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty