Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Rhodium(II) acetate dimer (∼46% Rh), anhydrous

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

AC26863-0010 AC26863-0050 AC26863-2500
TS26863-0010EA 1152 USD InStock
TS26863-0010 TS26863-0050 TS26863-2500
Rhodium(II) acetate dimer (∼46% Rh), anhydrous
Rhodium (II) acetate dimer


Formula: C₈H₁₂O₈Rh₂
MW: 441.987 g/mol
Melting Pt: 205 °C
Storage Temperature: Ambient
MDL Number: MFCD00003538
CAS Number: 15956-28-2
EINECS: 240-084-8
Order Now
Specification Test Results
Appearance Green Crystalline powder
Infrared spectrum Conforms
Assay 46 % (Rh)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty