Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane 96%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

AC21528-0050 AC21528-0250
TS21528-0050EA 159 USD InStock
TS21528-0050 TS21528-0250
1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane 96%
1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane


Formula: C₁₂H₂₈Cl₂OSi₂
MW: 315.43 g/mol
Boiling Pt: 120 °C (15 mmHg)
Density: 1 g/cm³
Flash Pt: 76 °C
Storage Temperature: Ambient
MDL Number: MFCD00009655
CAS Number: 69304-37-6
UN: 2987
ADR: 8 II
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4533 to 1.4553 (20°C, 589 nm)
GC ≥95 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty