Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Lanthanum(III) isopropoxide 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

AC38364-0010 AC38364-0050
TS38364-0010EA 104 USD InStock
TS38364-0010 TS38364-0050
Lanthanum(III) isopropoxide 98%
Lanthanum (III) isopropoxide


Formula: C₉H₂₁LaO₃
Storage Temperature: Ambient
MDL Number: MFCD00015305
CAS Number: 19446-52-7
UN: 2925
ADR: 4.1,II
Order Now
Specification Test Results
Appearance White to light yellow Powder
Infrared spectrum Conforms
Titration Complexometric ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty