Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluorphenylmagnesium bromide 2 mol/l in diethyl ether, AcroSeal™

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

AC37737-1000 AC37737-8000
TS37737-1000EA 91 USD InStock
TS37737-1000 TS37737-8000
4-Fluorphenylmagnesium bromide 2 mol/l in diethyl ether, AcroSeal™
4-Fluorphenylmagnesium bromide


Formula: C₆H₄BrFMg
Density: 1.043 g/cm³
Flash Pt: –40 °C
Storage Temperature: Ambient
MDL Number: MFCD00009667
CAS Number: 352-13-6
UN: 3399
ADR: 4.3,I
Order Now
Specification Test Results
Appearance Clear to turbid yellow to brown Solution (May develop some turbidity or precipitate)
Titration Grignard 1.9 to 2.2 M

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty