Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethoxymethylsilane 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

AC39226-0050 AC39226-0250 AC39226-1000
TS39226-0050EA 51 USD InStock
TS39226-0050 TS39226-0250 TS39226-1000
Diethoxymethylsilane 97%
Diethoxymethylsilane


Formula: C₅H₁₄O₂Si
Flash Pt: 10 °C
Storage Temperature: Ambient
MDL Number: MFCD00026749
CAS Number: 2031-62-1
UN: 1993
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3740 to 1.3760 (20°C, 589 nm)
GC ≥96.0 %

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty