Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(Methoxymethyl)-N-(trimethylsilylmethyl)benzylamine 96%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

AC34430-0050 AC34430-0250
TS34430-0050EA 120 USD InStock
TS34430-0050 TS34430-0250
N-(Methoxymethyl)-N-(trimethylsilylmethyl)benzylamine 96%
N-(Methoxymethyl)-N-(trimethylsilylmethyl)benzylamine


Formula: C₁₃H₂₃NOSi
MW: 237.42 g/mol
Boiling Pt: 76 °C (.3 mmHg)
Density: 0.928 g/cm³
Flash Pt: 66 °C
Storage Temperature: Ambient
MDL Number: MFCD00674005
CAS Number: 93102-05-7
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4910 to 1.4940 (20°C, 589 nm)
Elemental Analysis ≥95 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty