Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Pyridylacetic acid hydrochloride 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

AC13195-0050 AC13195-0250
TS13195-0050EA 68 USD InStock
TS13195-0050 TS13195-0250
2-Pyridylacetic acid hydrochloride 99%
2-Pyridylacetic acid hydrochloride


Formula: C₇H₇NO₂
Melting Pt: 135 °C
Storage Temperature: Ambient
MDL Number: MFCD00012812
CAS Number: 16179-97-8
Order Now
Specification Test Results
Appearance White to beige Crystalline powder or crystals
Infrared spectrum Conforms
Titration Argentometric ≥98.5 %
HPLC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty