Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bis[eta-(2,5-norbornadiene)]rhodium(I) tetrafluoroborate 94%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

AC38255-1000 AC38255-5000
TS38255-1000EA 90 USD InStock
TS38255-1000 TS38255-5000
Bis[eta-(2,5-norbornadiene)]rhodium(I) tetrafluoroborate 94%
Bis[eta-(2,5-norbornadiene)]rhodium(I) tetrafluoroborate


Formula: C₁₄H₈BF₄Rh
MW: 365.92 g/mol
Melting Pt: 209…211 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00671775
CAS Number: 36620-11-8
UN: 1325
ADR: 4.1 II
Order Now
Specification Test Results
Appearance Red Powder
Infrared spectrum Conforms
Elemental Analysis ≥92 %

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty