Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dicobalt octacarbonyl 95% stabilised

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

AC29184-0050 AC29184-0250 AC29184-1000
TS29184-0050EA 100.38 USD InStock
TS29184-0050 TS29184-0250 TS29184-1000
Dicobalt octacarbonyl 95% stabilised
Dicobalt octacarbonyl


Formula: C₈Co₂O₈
MW: 341.95 g/mol
Melting Pt: 51…52 °C (dec.)
Density: 1.81 g/cm³
Flash Pt: –13 °C
Storage Temperature: Freezer
MDL Number: MFCD00016024
CAS Number: 10210-68-1
UN: 3124
ADR: 6.1 I
Order Now
Specification Test Results
Appearance Orange-red to dark red Crystals or flakes
Infrared spectrum Conforms
Assay ≥94 %
Stabilizer ≤5 % hexane

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty