Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl mercaptan sodium salt 95%, pure

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

AC30571-0010 AC30571-0050 AC30571-0250 AC30571-1000
TS30571-0010EA 48 USD InStock
TS30571-0010 TS30571-0050 TS30571-0250 TS30571-1000
Methyl mercaptan sodium salt 95%, pure
Methyl mercaptan sodium salt


Formula: CH₃NaS
MW: 70.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00174316
CAS Number: 5188-07-8
UN: 2921
ADR: 8 I
Order Now
Specification Test Results
Appearance White to light beige Powder or granular powder
Titration Iodimetric 94 to 106 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty