Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methylmagnesium Iodide 3 M in diethyl ether, AcroSeal™

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

AC42740-1000 AC42740-8000
TS42740-1000EA 79 USD InStock
TS42740-1000 TS42740-8000
Methylmagnesium Iodide 3 M in diethyl ether, AcroSeal™
Methylmagnesium Iodide


Formula: CH₃IMg
Density: 1.26 g/cm³
Flash Pt: –40 °C
Storage Temperature: Ambient
MDL Number: MFCD00001026
CAS Number: 917-64-6
UN: 3399
ADR: 4.3,I
Order Now
Specification Test Results
Appearance Yellow to gray to brown Clear to hazy solution
Strength 2.85 to 3.50 M
Refractive index 1.4920 to 1.4940 (20°C, 589 nm)
GC ≥97.5 %

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty