Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

O,O'-Di-2-pyridyl thiocarbonate 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

AC39088-0010 AC39088-0050
TS39088-0010EA 91 USD InStock
TS39088-0010 TS39088-0050
O,O'-Di-2-pyridyl thiocarbonate 97%
O,O'-Di-2-pyridyl thiocarbonate


Formula: C₁₁H₈N₂O₂S
Melting Pt: 96…99 °C
Storage Temperature: Freezer
MDL Number: MFCD00074870
CAS Number: 96989-50-3
Order Now
Specification Test Results
Appearance Yellow to brown Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty