Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium diformylamide 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

AC38043-0050 AC38043-0250
TS38043-0050EA 65 USD InStock
TS38043-0050 TS38043-0250
Sodium diformylamide 98%
Sodium diformylamide


Formula: C₂H₃NNaO₂
MW: 96.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00145302
CAS Number: 18197-26-7
Order Now
Specification Test Results
Appearance Light beige Powder
Infrared spectrum Conforms
Titration with HCl ≥97.5 %

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty