Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tetrakis(acetonitrile)copper(I) tetrafluoroborate

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

AC39876-0010 AC39876-0050
TS39876-0010EA 32 USD InStock
TS39876-0010 TS39876-0050
Tetrakis(acetonitrile)copper(I) tetrafluoroborate
Tetrakis(acetonitrile)copper(I) tetrafluoroborate


Formula: C₈H₁₂BCuF₄N₄
MW: 314.56 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09265110
CAS Number: 15418-29-8
UN: 1759
ADR: 8 II
Order Now
Specification Test Results
Appearance White Crystalline powder
Infrared spectrum Conforms
Titration Complexometric 19.5 to 20.5 % Cu

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty