Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Trimethylboroxine 50% (w/w) 3.5 M in THF

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

AC29752-0250 AC29752-1000
TS29752-0250EA 254 USD InStock
TS29752-0250 TS29752-1000
Trimethylboroxine 50% (w/w) 3.5 M in THF
Trimethylboroxine


Formula: C₃H₉B₃O₃
Boiling Pt: 78…80 °C (760 mmHg)
Melting Pt: –38 °C
Density: 0.89 g/cm³
Flash Pt: –9 °C
Storage Temperature: Ambient
MDL Number: MFCD00013354
CAS Number: 823-96-1
UN: 1993
ADR: 3,II
Order Now
Specification Test Results
Appearance Colorless to yellow Solution
Assay 40.0 to 60.0 % (ex Boron)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty