Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

trans-Dichlorobis(triethylphosphine)palladium(II)

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

AC37797-0010 AC37797-2500
TS37797-0010EA 150 USD InStock
TS37797-0010 TS37797-2500
trans-Dichlorobis(triethylphosphine)palladium(II)
trans-Dichlorobis(triethylphosphine)palladium(II)


Formula: C₁₂H₃₂Cl₂P₂Pd
Melting Pt: 139…142 °C
Storage Temperature: Ambient
MDL Number: MFCD00191831
CAS Number: 28425-04-9
UN: 3464
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Yellow Powder
Infrared spectrum Conforms
Palladium (Pd) ≥24.9 %

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty