Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Magnesium bromide diethyl etherate 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

AC20970-0250 AC20970-1000
TS20970-0250EA 60 USD InStock
TS20970-0250 TS20970-1000
Magnesium bromide diethyl etherate 99%
Magnesium bromide diethyl etherate


Formula: C₄H₁₀Br₂MgO
Flash Pt: 35 °C
Storage Temperature: Ambient
MDL Number: MFCD00064500
CAS Number: 29858-07-9
UN: 1325
ADR: 4.1,II
Order Now
Specification Test Results
Appearance Off-white to beige Crystalline powder and/or chunks
Infrared spectrum Conforms
Titration Argentometric ≥98.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty