Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tetraethylammonium acetate tetrahydrate 97+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

AC19494-0250 AC19494-1000 AC19494-5000
TS19494-0250EA 94 USD InStock
TS19494-0250 TS19494-1000 TS19494-5000
Tetraethylammonium acetate tetrahydrate 97+%
Tetraethylammonium acetate tetrahydrate


Formula: C₁₀H₃₁NO₆
Storage Temperature: Ambient
MDL Number: MFCD00149993
CAS Number: 67533-12-4
UN: 1325
Order Now
Specification Test Results
Appearance White to light yellow Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 41 to 46 °C
Titration with HClO4 97.0 to 103.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty