Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Phenethylmagnesium chloride solution 1 M in THF, AcroSeal™

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

AC42780-1000 AC42780-8000
TS42780-1000EA 72 USD InStock
TS42780-1000 TS42780-8000
Phenethylmagnesium chloride solution 1 M in THF, AcroSeal™
Phenethylmagnesium chloride solution


Formula: C₈H₉ClMg
Density: 0.954 g/cm³
Flash Pt: –17 °C
Storage Temperature: Ambient
CAS Number: 90878-19-6
UN: 2924
ADR: 3,II
Order Now
Specification Test Results
Appearance Yellow to brown Solution
Titration Grignard 0.90 to 1.10 M
Refractive index 1.3820 to 1.3940 (20°C, 589 nm)
GC ≥98.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty