Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Carbonylchlorohydridotris(triphenylphosphine)ruthenium(II)

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

AC39214-0010 AC39214-0050
TS39214-0010EA 114 USD InStock
TS39214-0010 TS39214-0050
Carbonylchlorohydridotris(triphenylphosphine)ruthenium(II)
Carbonylchlorohydridotris(triphenylphosphine)ruthenium(II)


Formula: C₅₅H₄₅ClOP₃Ru
Melting Pt: 204…206 °C
Storage Temperature: Ambient
MDL Number: MFCD00049804
CAS Number: 16971-33-8
Order Now
Specification Test Results
Appearance Cream to tan to brown Powder or crystals
Infrared spectrum Conforms
Ruthenium (Ru) ≥10.3 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty