Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethylenebis(triphenylphosphine)platinum(0) 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

AC36936-0010 AC36936-2500
TS36936-0010EA 415 USD InStock
TS36936-0010 TS36936-2500
Ethylenebis(triphenylphosphine)platinum(0) 98%
Ethylenebis(triphenylphosphine)platinum(0)


Formula: C₃₈H₃₄P₂Pt
Melting Pt: 127…131 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00012028
CAS Number: 12120-15-9
Order Now
Specification Test Results
Appearance Light yellow to beige Powder
Infrared spectrum Conforms
Melting point 127 to 131 °C
Assay ≥97.5 % (ex Pt)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty