Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methyl-1-propenylmagnesium bromide 0.5 M in THF, AcroSeal™

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

AC43556-1000 AC43556-8000
TS43556-1000EA 145 USD InStock
TS43556-1000 TS43556-8000
2-Methyl-1-propenylmagnesium bromide 0.5 M in THF, AcroSeal™
2-Methyl-1-propenylmagnesium bromide


Formula: C₄H₇BrMg
Density: 0.952 g/cm³
Flash Pt: –15 °C
Storage Temperature: Ambient
MDL Number: MFCD00209600
CAS Number: 38614-36-7
UN: 3399
ADR: 4.3,I
Order Now
Specification Test Results
Appearance Yellow to orange Solution
Titration Grignard 0.45 to 0.65 M
Refractive index 1.3390 to 1.3410 (20°C, 589 nm)
GC ≥98.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty