Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methyl-2-phenylpropylmagnesium chloride 0.5 M in diethyl ether, AcroSeal™

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

AC43875-1000 AC43875-8000
TS43875-1000EA 95 USD InStock
TS43875-1000 TS43875-8000
2-Methyl-2-phenylpropylmagnesium chloride 0.5 M in diethyl ether, AcroSeal™
2-Methyl-2-phenylpropylmagnesium chloride


Formula: C₁₀H₁₃ClMg
MW: 192.97 g/mol
Density: 0.76 g/cm³
Flash Pt: –28 °C
Storage Temperature: Ambient
MDL Number: MFCD00075586
CAS Number: 35293-35-7
UN: 2924
ADR: 3 I
Order Now
Specification Test Results
Appearance Colorless to yellow to gray Solution (May develop some turbidity or precipitate)
Titration Active base 0.40 to 0.60 M

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty