Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium trimethylsilanolate 2 M in THF, AcroSeal™

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

AC44846-1000 AC44846-8000
TS44846-1000EA 102 USD InStock
TS44846-1000 TS44846-8000
Potassium trimethylsilanolate 2 M in THF, AcroSeal™
Potassium trimethylsilanolate


Formula: C₃H₉KOSi
Density: 0.91 g/cm³
Flash Pt: –17 °C
Storage Temperature: Ambient
MDL Number: MFCD00013421
CAS Number: 10519-96-7
UN: 2924
ADR: 3,II
Order Now
Specification Test Results
Appearance Colorless to straw yellow Solution
Titration with HClO4 1.9 to 2.2 M

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty