Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tris(acetonitrile)cyclopentadienylruthenium(II) hexafluorophosphate 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

AC37789-0010 AC37789-2500
TS37789-0010EA 488 USD InStock
TS37789-0010 TS37789-2500
Tris(acetonitrile)cyclopentadienylruthenium(II) hexafluorophosphate 98%
Tris(acetonitrile)cyclopentadienylruthenium(II) hexafluorophosphate


Formula: C₁₁H₉F₆N₃PRu
Storage Temperature: Refrigerator
MDL Number: MFCD02684569
CAS Number: 80049-61-2
Order Now
Specification Test Results
Appearance Yellow to orange to brown Powder
Infrared spectrum Conforms
Assay ≥97.5 % (ex Ru)

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty